Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-2-methylpentane 98.0+%, TCI America™
SDP

Catalog No. C14765G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Catalog No. C14765G

Supplier: TCI America C14765G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14753-05-0
Molecular Formula C6H13Cl
Molecular Weight (g/mol) 120.62
MDL Number MFCD00053236
InChI Key SLBTUZZYJLBZQV-UHFFFAOYSA-N
PubChem CID 527856
IUPAC Name 1-chloro-2-methylpentane
SMILES CCCC(C)CCl

Specifications

Color Colorless
Boiling Point 120°C
Quantity 5 g
UN Number 1993
Formula Weight 120.62
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-2-methylpentane
EINECSNumber (2)-0066
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.