Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Benzyl-5-oxopyrrolidine-3-carboxylic Acid 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
1 g
Specifications
Specifications
| Melting Point | 144°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 219.24 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-Benzyl-5-oxopyrrolidine-3-carboxylic Acid |
Safety and Handling
| RTECSNumber | UY0686500 |
| TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction