Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(5-Fluoro-2,4-dinitrophenyl)-4-methylpiperazine 98.0+%, TCI America™
SDP

Catalog No. F0650100MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g

Catalog No. F0650100MG

Supplier: TCI America F0650100MG

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 928830-73-3
Molecular Formula C11H13FN4O4
Molecular Weight (g/mol) 284.25
MDL Number MFCD10000959
InChI Key SSBAFRIGYXRBRK-UHFFFAOYSA-N
Synonym PPZ
PubChem CID 16223480
IUPAC Name 1-(5-fluoro-2,4-dinitrophenyl)-4-methylpiperazine
SMILES CN1CCN(CC1)C1=CC(F)=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 115°C
Color Yellow
Quantity 100 mg
Formula Weight 284.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(5-Fluoro-2,4-dinitrophenyl)-4-methylpiperazine
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.