Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dibromo-2,2-dimethoxypropane 98.0+%, TCI America™
SDP

Catalog No. D45795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45795G 5 g
D457925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45795G Supplier TCI America Supplier No. D45795G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22094-18-4
Molecular Formula C5H10Br2O2
Molecular Weight (g/mol) 261.941
MDL Number MFCD00051792
InChI Key CPAHOXOBYHMHDT-UHFFFAOYSA-N
Synonym 1,3-dibromoacetone dimethyl acetal, 1,3-acetone dibromide dimethyl acetal, 1,3-dibromo-2,2-dimethoxy-propane, 2-bromo-1-bromomethyl-1,1-dimethoxyethane, 1,3-dibromoacetone dimethyl ketal, propane, 1,3-dibromo-2,2-dimethoxy, pubchem2043, ag120 intermediates, acmc-1ck33, timtec-bb sbb039320
PubChem CID 2734196
IUPAC Name 1,3-dibromo-2,2-dimethoxypropane
SMILES COC(CBr)(CBr)OC

Specifications

Melting Point 65°C
Quantity 5 g
Formula Weight 261.94
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dibromo-2,2-dimethoxypropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.