Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzothiazole-2-carbaldehyde, Thermo Scientific™

Catalog No. CC06004CB
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC06004CB 250 mg
CC06004DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC06004CB Supplier Thermo Scientific Chemicals Supplier No. CC06004CB
Only null left
Add to Cart

Chemical Identifiers

CAS 6639-57-2
Molecular Formula C8H5NOS
Molecular Weight (g/mol) 163.194
MDL Number MFCD00526215
InChI Key RHKPJTFLRQNNGJ-UHFFFAOYSA-N
Synonym 2-benzothiazolecarboxaldehyde, benzothiazole-2-carbaldehyde, benzothiazole-2-carboxaldehyde, benzo d thiazole-2-carbaldehyde, 1,3-benzothiazole-2-carboxaldehyde, 2-formylbenzothiazole, zlchem 104, benzothiazolecarbaldehyde, pubchem14482, 2-benzothiazolecarbaldehyde
PubChem CID 241608
IUPAC Name 1,3-benzothiazole-2-carbaldehyde
SMILES C1=CC=C2C(=C1)N=C(S2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 163.2
Percent Purity 97%
Chemical Name or Material 1, 3-Benzothiazole-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.