Learn More
1,2-Dibromoethane, 98%
Description
1,2-dibromoethane is widely used as a bromine source in organic synthesis. It is utilized as a fumigant for treatment of logs for termites and beetles, for control of moths in beehives. It finds the application to make vinyl bromide, a precursor to some fire retardants. It is used as an intermediate in the preparation of dyes and pharmaceuticals. It is also used as a lead scavenger in antiknock mixtures added to gasolines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,2-dibromoethane is widely used as a bromine source in organic synthesis. It is utilized as a fumigant for treatment of logs for termites and beetles, for control of moths in beehives. It finds the application to make vinyl bromide, a precursor to some fire retardants. It is used as an intermediate in the preparation of dyes and pharmaceuticals. It is also used as a lead scavenger in antiknock mixtures added to gasolines.
Solubility
Miscible with alcohol, acetone, benzene, diethyl ether and ethyl acetate. Slightly miscible with water.
Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com.
Specifications
Specifications
| Melting Point | 9°C to 10°C |
| Density | 2.179 |
| Boiling Point | 132°C |
| Flash Point | None |
| Refractive Index | 1.5385 |
| Linear Formula | BrCH2CH2Br |
| Quantity | 250 g |
| UN Number | UN1605 |
| Sensitivity | Light sensitive |
| Merck Index | 14,3796 |
| Show More |
Safety and Handling
| Hazard Category | H301+H311-H315-H319-H335-H350 |
| Hazard Statement | GHS H Statement H301-H311-H331-H350-H315-H319-H335 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. May cause cancer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ETHYLENE DIBROMIDE |
| EINECSNumber | 203-444-5 |
| RTECSNumber | KH9275000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.