Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Di-n-octyl-4,4'-bipyridinium Dibromide 98.0+%, TCI America™
SDP

Catalog No. d18545g Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Catalog No. D18545G

Supplier: TCI America D18545G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 36437-30-6
Molecular Formula C26H42Br2N2
Molecular Weight (g/mol) 542.44
MDL Number MFCD00060051
InChI Key GVUBEJANEHZYPA-UHFFFAOYSA-L
PubChem CID 37451
IUPAC Name 1,1'-dioctyl-[4,4'-bipyridine]-1,1'-diium dibromide
SMILES [Br-].[Br-].CCCCCCCC[N+]1=CC=C(C=C1)C1=CC=[N+](CCCCCCCC)C=C1

Specifications

pH 5.0 to 7.0 (25g/L H2O soln.)
Quantity 5 g
Formula Weight 542.44
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Di-n-octyl-4,4′-bipyridinium Dibromide
RTECSNumber DW2380000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.