Learn More
Sartorius Tacta™ Mechanical Pipettes
GREENER_CHOICE
Exceptionally balanced design is effortless to use and exceptionally comfortable to hold
Supplier:  Sartorius LH729070
 View more versions of this product
Catalog No. 14-577-798
Add to Cart
Description
Description
  • Comfortable to hold due to the ergonomically designed handle
  • Unique Sartorius Optilock system provides flexibility for volume adjustment and locking
  • Controlled and smooth tip ejection with the new Sartorius Optiject technology
  • Calibration adjustment to provide accurate results for various liquid types
  • Easy to clean, with only three parts to disassemble
  • Reliable results, even over lengthy pipetting periods
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Optifit Tips: 1000μL, SafetySpace Filter Tips: 1000μL
Blue
Tacta™
100 to 1000 μL
Autoclavable
1
Pipette
Yellow
Product Suggestions
Product Suggestions