Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrrole 99.0+%, TCI America™
SDP

Catalog No. P057425ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL

Catalog No. P057425ML

Supplier: TCI America P057425ML

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 109-97-7
Molecular Formula C4H5N
Molecular Weight (g/mol) 67.09
MDL Number MFCD00005216
InChI Key KAESVJOAVNADME-UHFFFAOYSA-N
Synonym pyrrole, divinylenimine, azole, imidole, pyrrol, monopyrrole, divinyleneimine, 1-aza-2,4-cyclopentadiene, polypyrrole, unii-86s1zd6l2c
PubChem CID 8027
ChEBI CHEBI:19203
IUPAC Name 1H-pyrrole
SMILES N1C=CC=C1

Specifications

Melting Point -23°C
Color Yellow
Boiling Point 130°C
Quantity 25 mL
UN Number 1992
Formula Weight 67.09
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Pyrrole
EINECSNumber (5)-0100
RTECSNumber UX9275000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.