Learn More
Invitrogen™ DNA-free™ DNA Removal Kit
Description
DNA-free™ DNase treatment and removal reagents are designed for removal of contaminating DNA from RNA samples and for removal of DNase after treatment.
- No organic extraction or heat inactivation required
- Includes novel reagent to remove DNase
- Recombinant DNase I is certified RNase-freeThe DNA-free
- Inactivation and removal of DNase
- DNA-free reagents effectively remove DNase and divalent cations from reaction mixture
- DNase/cation removal step takes only three minutes
- No organic extraction, EDTA addition, or heat inactivation is required
TURBO™ DNA-free™ Kit is similar to the DNA-free Kit but includes TURBO DNase, an engineered hyperactive DNase that exhibits up to 350% greater catalytic efficiency than wild type DNase. The enzyme also has a 6-fold lower Km for DNA, thus enabling effective removal of trace quantities of DNA contamination.
PCR & Real-Time PCR, Real Time PCR (qPCR), Reverse Transcription
Specifications
Specifications
| Content And Storage | Store rDNase1 , 10X DNase 1 Buffer and DNase Inactivation Reagent at -20°C. Store Nuclease–free Water at room temperature. |
| Shipping Condition | Dry Ice |
| Enzyme | DNase |
| Compatible Buffer | 10X Reaction Buffer |
| Quantity | 50 reactions |
| Product Type | DNA Removal Kit |
| Product Line | Ambion, DNA-free |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.