For security reasons you will be automatically logged out after 30 minutes of inactivity.
Denver Instrument* Pinnacle* Series Analytical Balances
Pinnacle Analytical Balance
Weighing capacities up to 310g
Simple, menu-driven operation allows quick access to powerful functions: multiple weighing units, parts counting, percent weighing, animal weighing. Built-in bidirectional RS-232-C interface and USB ports enable a network of analytical laboratory instruments centered on your P-Series balance. View your balance anywhere on the network via Ethernet.
See details